Assay Details
| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027740.6 | NP_082016.1 | 2 - 3 | 140 | 72 | |
| GenBank mRNA | AK016991.1 | - | 1 - 2 | 64 | 72 | |
| AK030180.1 | - | 1 - 2 | 34 | 72 | ||
| AK045302.1 | - | 2 - 3 | 119 | 72 | ||
| AK050233.1 | - | 2 - 3 | 209 | 72 | ||
| AK090075.1 | - | 2 - 3 | 143 | 72 | ||
| BC050888.1 | - | 1 - 2 | 138 | 72 | ||
| BC099926.1 | - | 2 - 3 | 227 | 72 | ||
| BC144819.1 | - | 2 - 3 | 229 | 72 | ||
| BC145801.1 | - | 2 - 3 | 229 | 40131104 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016991.1, AK030180.1 transcripts.
Set Membership: |
|

Poc1b
Genomic Map