Assay Details
| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027740.6 | NP_082016.1 | 4 - 5 | 493 | 77 | |
| GenBank mRNA | AK016991.1 | - | 3 - 4 | 417 | 77 | |
| AK030180.1 | - | 3 - 4 | 387 | 77 | ||
| AK045302.1 | - | 4 - 5 | 472 | 77 | ||
| AK050233.1 | - | 4 - 5 | 562 | 77 | ||
| AK090075.1 | - | 4 - 5 | 496 | 77 | ||
| BC050888.1 | - | 3 - 4 | 491 | 77 | ||
| BC099926.1 | - | 4 - 5 | 580 | 77 | ||
| BC144819.1 | - | 4 - 5 | 582 | 77 | ||
| BC145801.1 | - | 4 - 5 | 582 | 40131104 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poc1b
Genomic Map