Assay Details
| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027740.6 | NP_082016.1 | 8 - 9 | 914 | 75 | |
| GenBank mRNA | AK016991.1 | - | 7 - 8 | 838 | 75 | |
| AK030180.1 | - | 7 - 8 | 808 | 75 | ||
| AK045302.1 | - | 8 - 9 | 893 | 75 | ||
| AK050233.1 | - | 8 - 9 | 983 | 75 | ||
| AK090075.1 | - | 8 - 9 | 917 | 75 | ||
| BC050888.1 | - | 7 - 8 | 912 | 75 | ||
| BC099926.1 | - | 8 - 9 | 1001 | 75 | ||
| BC144819.1 | - | 8 - 9 | 1003 | 75 | ||
| BC145801.1 | - | 8 - 9 | 1003 | 40131104 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poc1b
Genomic Map