Assay Details
| Gene Symbol: Fam65c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080708.2 | NP_001074177.1 | 19 - 20 | 2626 | 72 | |
| GenBank mRNA | AK158217.1 | - | 19 - 20 | 2570 | 72 | |
| BC130272.1 | - | 19 - 20 | 2626 | 40143210 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam65c
Genomic Map