Assay Details
| Gene Symbol: Def8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253783.1 | NP_001240712.1 | 11 - 12 | 1206 | 67 | |
| NM_001253784.1 | NP_001240713.1 | 10 - 11 | 1184 | 67 | ||
| NM_001281803.1 | NP_001268732.1 | 9 - 10 | 1188 | 67 | ||
| NM_054046.5 | NP_473387.1 | 9 - 10 | 1097 | 67 | ||
| GenBank mRNA | AK028726.1 | - | 10 - 11 | 1156 | 67 | |
| AK034028.1 | - | 1063 | 67 | |||
| AK046271.1 | - | 9 - 10 | 1057 | 67 | ||
| AK050405.1 | - | 10 - 11 | 1155 | 67 | ||
| AK081295.1 | - | 9 - 10 | 1087 | 67 | ||
| AK086174.1 | - | 9 - 10 | 1060 | 67 | ||
| AK087778.1 | - | 9 - 10 | 1068 | 67 | ||
| AK129019.1 | - | 9 - 10 | 1139 | 67 | ||
| AK133979.1 | - | 9 - 10 | 1071 | 67 | ||
| AK150334.1 | - | 9 - 10 | 1033 | 67 | ||
| AK150994.1 | - | 9 - 10 | 1033 | 67 | ||
| AK164147.1 | - | 9 - 10 | 1069 | 67 | ||
| BC003306.1 | - | 11 - 12 | 1157 | 3154712 | 67 | |
| CN705834.1 | - | 2 - 3 | 138 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Def8
Genomic Map