Assay Details
| Gene Symbol: Slc7a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178798.3 | NP_848913.1 | 5 - 6 | 1034 | 95 | |
| GenBank mRNA | AK030762.1 | - | 6 - 7 | 836 | 95 | |
| AK046548.1 | - | 4 - 5 | 1701 | 95 | ||
| AK049478.1 | - | 5 - 6 | 1034 | 95 | ||
| AK053899.1 | - | 5 - 6 | 1043 | 95 | ||
| AK088592.1 | - | 5 - 6 | 951 | 95 | ||
| AK129099.1 | - | 2 - 3 | 2430 | 95 | ||
| AK157127.1 | - | 5 - 6 | 1173 | 95 | ||
| AK157379.1 | - | 4 - 5 | 1703 | 95 | ||
| AK164666.1 | - | 5 - 6 | 1035 | 95 | ||
| AK165629.1 | - | 5 - 6 | 1051 | 95 | ||
| AK165979.1 | - | 5 - 6 | 1053 | 95 | ||
| BC038404.1 | - | 4 - 5 | 968 | 4949962 | 95 | |
| BC042898.1 | - | 2 - 3 | 1566 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a6
Genomic Map