Assay Details
| Gene Symbol: Slc7a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178798.3 | NP_848913.1 | 6 - 7 | 1142 | 99 | |
| GenBank mRNA | AK030762.1 | - | 7 - 8 | 944 | 99 | |
| AK046548.1 | - | 5 - 6 | 1809 | 99 | ||
| AK049478.1 | - | 6 - 7 | 1142 | 99 | ||
| AK053899.1 | - | 6 - 7 | 1151 | 99 | ||
| AK088592.1 | - | 6 - 7 | 1059 | 99 | ||
| AK129099.1 | - | 3 - 4 | 2538 | 99 | ||
| AK157127.1 | - | 6 - 7 | 1281 | 99 | ||
| AK157379.1 | - | 5 - 6 | 1811 | 99 | ||
| AK164666.1 | - | 6 - 7 | 1143 | 99 | ||
| AK165629.1 | - | 6 - 7 | 1159 | 99 | ||
| AK165979.1 | - | 6 - 7 | 1161 | 99 | ||
| BC042898.1 | - | 3 - 4 | 1674 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a6
Genomic Map