| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800009324.mm.1 |
NM_001163756.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009324.mm.1 |
NM_001163756.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009324.mm.1 |
NM_172759.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009324.mm.1 |
NM_172759.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009324.mm.1 |
TC0800001196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800009322.mm.1 |
NM_001163756.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009322.mm.1 |
NM_001163756.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009322.mm.1 |
NM_172759.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009322.mm.1 |
NM_172759.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009322.mm.1 |
TC0800001196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800009323.mm.1 |
NM_001163756.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009323.mm.1 |
NM_001163756.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009323.mm.1 |
NM_172759.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009323.mm.1 |
NM_172759.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009323.mm.1 |
TC0800001196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800009326.mm.1 |
NM_001163756.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009326.mm.1 |
NM_001163756.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009326.mm.1 |
NM_172759.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009326.mm.1 |
NM_172759.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009326.mm.1 |
TC0800001196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800009325.mm.1 |
NM_001163756.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009325.mm.1 |
NM_001163756.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009325.mm.1 |
NM_172759.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009325.mm.1 |
NM_172759.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009325.mm.1 |
TC0800001196.mm.1 |
MTA TC |