Assay Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081241.2 | NP_001074710.2 | 12 - 13 | 1228 | 74 | |
| GenBank mRNA | AK052316.1 | - | 12 - 13 | 1177 | 74 | |
| AK122573.1 | - | 12 - 13 | 1202 | 74 | ||
| BC030451.1 | - | 2 - 3 | 291 | 5400266 | 74 | |
| BC079880.1 | - | 12 - 13 | 1142 | 6852251 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC030451.1 transcripts.
Set Membership: |
|

Fam65a
Genomic Map