Assay Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081241.2 | NP_001074710.2 | 17 - 18 | 3189 | 55 | |
| GenBank mRNA | AK052316.1 | - | 18 - 19 | 3070 | 55 | |
| AK122573.1 | - | 17 - 18 | 3163 | 55 | ||
| BC030451.1 | - | 7 - 8 | 2252 | 5400266 | 55 | |
| BC079880.1 | - | 17 - 18 | 3103 | 6852251 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam65a
Genomic Map