Assay Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081241.2 | NP_001074710.2 | 3 - 4 | 398 | 60 | |
| GenBank mRNA | AK052316.1 | - | 3 - 4 | 347 | 60 | |
| AK122573.1 | - | 3 - 4 | 372 | 60 | ||
| BB852730.1 | - | 3 - 4 | 397 | 60 | ||
| BC079880.1 | - | 3 - 4 | 312 | 6852251 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam65a
Genomic Map