Assay Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081241.2 | NP_001074710.2 | 6 - 7 | 565 | 68 | |
| GenBank mRNA | AK052316.1 | - | 6 - 7 | 514 | 68 | |
| AK122573.1 | - | 6 - 7 | 539 | 68 | ||
| AK186816.1 | - | 2 - 3 | 90 | 68 | ||
| BC079880.1 | - | 6 - 7 | 479 | 6852251 | 68 | |
More Information
Set Membership: |
|

Fam65a
Genomic Map