Assay Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081241.2 | NP_001074710.2 | 7 - 8 | 716 | 93 | |
| GenBank mRNA | AK052316.1 | - | 7 - 8 | 665 | 93 | |
| AK122573.1 | - | 7 - 8 | 690 | 93 | ||
| AK186816.1 | - | 3 - 4 | 241 | 93 | ||
| BC079880.1 | - | 7 - 8 | 630 | 6852251 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam65a
Genomic Map