Assay Details
| Gene Symbol: Ednra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010332.2 | NP_034462.1 | 1 - 2 | 354 | 63 | |
| GenBank mRNA | AK042211.1 | - | 1 - 2 | 368 | 63 | |
| AK043210.1 | - | 1 - 2 | 251 | 63 | ||
| AK135555.1 | - | 1 - 2 | 247 | 63 | ||
| BC008277.1 | - | 1 - 2 | 338 | 2812426 | 63 | |
| CN669253.1 | - | 1 - 2 | 354 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK043210.1, BC008277.1, NM_010332.2 transcripts.
Set Membership: |
|

Ednra
Genomic Map