Assay Details
| Gene Symbol: Ednra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010332.2 | NP_034462.1 | 6 - 7 | 1444 | 71 | |
| GenBank mRNA | AF039892.1 | - | 5 - 6 | 739 | 71 | |
| AK043210.1 | - | 6 - 7 | 1341 | 71 | ||
| AK135555.1 | - | 6 - 7 | 1338 | 71 | ||
| BC008277.1 | - | 6 - 7 | 1428 | 2812426 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ednra
Genomic Map