Assay Details
| Gene Symbol: Casp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078836.1 | - | 1 - 2 | 56 | 75 | |
| AK151547.1 | - | 1 - 2 | 55 | 75 | ||
| AK156667.1 | - | 1 - 2 | 50 | 75 | ||
| AK160328.1 | - | 1 - 2 | 50 | 75 | ||
| AK171877.1 | - | 1 - 2 | 47 | 75 | ||
| BC061255.1 | - | 1 - 2 | 54 | 30251263 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151547.1, AK171877.1, BC061255.1 transcripts.
Set Membership: |
|

Casp4
Genomic Map