Assay Details
| Gene Symbol: Casp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138730.1 | - | 3 - 4 | 240 | 62 | |
| AK151547.1 | - | 3 - 4 | 408 | 62 | ||
| AK156667.1 | - | 3 - 4 | 403 | 62 | ||
| AK160328.1 | - | 3 - 4 | 403 | 62 | ||
| AK171877.1 | - | 3 - 4 | 400 | 62 | ||
| U59463.1 | - | 3 - 4 | 393 | 62 | ||
| Y13089.1 | - | 3 - 4 | 385 | 62 | ||
More Information
Set Membership: |
|

Casp4
Genomic Map