Assay Details
| Gene Symbol: Casp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB480706.1 | - | 3 - 4 | 670 | 84 | |
| AK078836.1 | - | 4 - 5 | 720 | 84 | ||
| AK138730.1 | - | 5 - 6 | 649 | 84 | ||
| AK151547.1 | - | 5 - 6 | 817 | 84 | ||
| AK156667.1 | - | 5 - 6 | 813 | 84 | ||
| AK171877.1 | - | 5 - 6 | 809 | 84 | ||
| BC061255.1 | - | 5 - 6 | 816 | 30251263 | 84 | |
| U59463.1 | - | 5 - 6 | 802 | 84 | ||
| Y13089.1 | - | 5 - 6 | 794 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp4
Genomic Map