Assay Details
| Gene Symbol: Ndrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195006.1 | NP_001181935.1 | 5 - 6 | 496 | 98 | |
| NM_145602.3 | NP_663577.1 | 5 - 6 | 496 | 98 | ||
| GenBank mRNA | AK090374.1 | - | 5 - 6 | 478 | 98 | |
| AK144294.1 | - | 5 - 6 | 478 | 98 | ||
| AK168873.1 | - | 5 - 6 | 480 | 98 | ||
| BC006595.1 | - | 5 - 6 | 470 | 3156802 | 98 | |
| BU059039.1 | - | 1 - 2 | 33 | 98 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK129305.1 | - | 791 | Chr. - - [-] on Build | 98 | ||
More Information
Set Membership: |
|

Ndrg4
Genomic Map