Assay Details
| Gene Symbol: A530013C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110334.1 | 1 - 2 | 42 | 59 | ||
| GenBank mRNA | AK166336.1 | - | 1 - 2 | 43 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P2160 | AK040674.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P2160 | AK040674.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P2160 | AK040674.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P2160 | AK040674.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2160 | AK040674.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2160 | AK040674.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01504301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01516400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01582249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02119124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02261911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21836966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02051777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21836970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21836973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21836975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02046306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01660028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01184715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01582242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01716573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01197415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01516410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01262121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01329856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01591353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01359214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01247812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01359212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01329855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01582245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02051775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01911708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01149980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01297698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01364033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21837156 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0200021385.mm.1 | NR_015500.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021385.mm.1 | NR_110334.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021385.mm.1 | TC0200002595.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200021380.mm.1 | NR_110334.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021380.mm.1 | TC0200002595.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200021384.mm.1 | NR_015500.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021384.mm.1 | NR_110334.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021384.mm.1 | TC0200002595.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 677812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3990097 | - | - |

A530013C23Rik
Genomic Map