Assay Details
| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178086.3 | NP_835187.2 | 2 - 3 | 434 | 117 | |
| GenBank mRNA | AK033736.1 | - | 2 - 3 | 432 | 117 | |
| AK090338.1 | - | 2 - 3 | 428 | 117 | ||
| AK161502.1 | - | 2 - 3 | 432 | 117 | ||
| AY660882.1 | - | 2 - 3 | 365 | 117 | ||
| BC026400.1 | - | 1 - 2 | 86 | 117 | ||
| BC046985.1 | - | 2 - 3 | 390 | 117 | ||
| BC111912.1 | - | 3 - 4 | 255 | 117 | ||
| BC128080.1 | - | 2 - 3 | 415 | 40104374 | 117 | |
| BC128081.1 | - | 2 - 3 | 415 | 40104375 | 117 | |
More Information
Set Membership: |
|

Fa2h
Genomic Map