Assay Details
| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178086.3 | NP_835187.2 | 4 - 5 | 680 | 63 | |
| GenBank mRNA | AK033736.1 | - | 4 - 5 | 678 | 63 | |
| AK090338.1 | - | 4 - 5 | 674 | 63 | ||
| AK161502.1 | - | 4 - 5 | 678 | 63 | ||
| AY660882.1 | - | 4 - 5 | 611 | 63 | ||
| BC026400.1 | - | 3 - 4 | 332 | 63 | ||
| BC026629.1 | - | 2 - 3 | 222 | 63 | ||
| BC046985.1 | - | 4 - 5 | 636 | 63 | ||
| BC111912.1 | - | 5 - 6 | 501 | 63 | ||
| BC128080.1 | - | 4 - 5 | 661 | 40104374 | 63 | |
| BC128081.1 | - | 4 - 5 | 661 | 40104375 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fa2h
Genomic Map