Assay Details
| Gene Symbol: Cog4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133973.2 | NP_598734.1 | 3 - 4 | 383 | 60 | |
| GenBank mRNA | AK156466.1 | - | 3 - 4 | 374 | 60 | |
| AK172465.1 | - | 3 - 4 | 374 | 60 | ||
| AK198969.1 | - | 1 - 2 | 36 | 60 | ||
| AK203917.1 | - | 1 - 2 | 36 | 60 | ||
| AY223866.1 | - | 3 - 4 | 410 | 60 | ||
| BC023120.1 | - | 3 - 4 | 360 | 4161858 | 60 | |
| CN843612.1 | - | 3 - 4 | 402 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cog4
Genomic Map