Assay Details
| Gene Symbol: Mtss1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198625.1 | NP_941027.1 | 12 - 13 | 1437 | 75 | |
| GenBank mRNA | AK052172.1 | - | 2 - 3 | 258 | 75 | |
| AK137832.1 | - | 12 - 13 | 1421 | 75 | ||
| BC060632.1 | - | 12 - 13 | 1437 | 6848397 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtss1l
Genomic Map