Assay Details
| Gene Symbol: Adgrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198894.1 | NP_001185823.1 | 1 - 2 | 309 | 60 | |
| GenBank mRNA | AF166382.1 | - | 1 - 2 | 309 | 60 | |
| AK085664.1 | - | 1 - 2 | 113 | 60 | ||
| AK087268.1 | - | 1 - 2 | 61 | 60 | ||
| AK164776.1 | - | 1 - 2 | 83 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF166382.1, AK087268.1, NM_001198894.1 transcripts.
Set Membership: |
|

Adgrg1
Genomic Map