Assay Details
| Gene Symbol: Adgrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198894.1 | NP_001185823.1 | 4 - 5 | 944 | 93 | |
| NM_018882.3 | NP_061370.2 | 4 - 5 | 894 | 93 | ||
| GenBank mRNA | AF166382.1 | - | 4 - 5 | 944 | 93 | |
| AK087268.1 | - | 4 - 5 | 696 | 93 | ||
| AK133678.1 | - | 4 - 5 | 661 | 93 | ||
| AK141886.1 | - | 4 - 5 | 923 | 93 | ||
| AK141894.1 | - | 4 - 5 | 707 | 93 | ||
| AK164776.1 | - | 4 - 5 | 719 | 93 | ||
| AK167547.1 | - | 3 - 4 | 648 | 93 | ||
| BC034678.1 | - | 4 - 5 | 690 | 3709247 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrg1
Genomic Map