Assay Details
| Gene Symbol: Adgrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198894.1 | NP_001185823.1 | 6 - 7 | 1225 | 61 | |
| NM_018882.3 | NP_061370.2 | 6 - 7 | 1175 | 61 | ||
| GenBank mRNA | AF166382.1 | - | 6 - 7 | 1225 | 61 | |
| AK087268.1 | - | 6 - 7 | 977 | 61 | ||
| AK133678.1 | - | 6 - 7 | 942 | 61 | ||
| AK141886.1 | - | 6 - 7 | 1204 | 61 | ||
| AK141894.1 | - | 6 - 7 | 988 | 61 | ||
| AK164776.1 | - | 6 - 7 | 1000 | 61 | ||
| AK167547.1 | - | 5 - 6 | 929 | 61 | ||
| BC034678.1 | - | 6 - 7 | 971 | 3709247 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrg1
Genomic Map