Assay Details
| Gene Symbol: Ciapin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134141.4 | NP_598902.1 | 2 - 3 | 280 | 76 | |
| GenBank mRNA | AK076116.1 | - | 2 - 3 | 236 | 76 | |
| AK079615.1 | - | 2 - 3 | 242 | 76 | ||
| AK088757.1 | - | 1 - 2 | 207 | 76 | ||
| AK136821.1 | - | 2 - 3 | 274 | 76 | ||
| AK146282.1 | - | 2 - 3 | 228 | 76 | ||
| AK163705.1 | - | 2 - 3 | 259 | 76 | ||
| AK168252.1 | - | 2 - 3 | 230 | 76 | ||
| AY523555.1 | - | 1 - 2 | 201 | 76 | ||
| BC016261.1 | - | 1 - 2 | 198 | 4920560 | 76 | |
| BC021864.1 | - | 2 - 3 | 220 | 5123958 | 76 | |
| BY283649.1 | - | 2 - 3 | 299 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ciapin1
Genomic Map