Assay Details
| Gene Symbol: Ciapin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134141.4 | NP_598902.1 | 5 - 6 | 666 | 63 | |
| GenBank mRNA | AK076116.1 | - | 5 - 6 | 622 | 63 | |
| AK079615.1 | - | 5 - 6 | 628 | 63 | ||
| AK088757.1 | - | 4 - 5 | 593 | 63 | ||
| AK136821.1 | - | 5 - 6 | 660 | 63 | ||
| AK163705.1 | - | 5 - 6 | 645 | 63 | ||
| AK168252.1 | - | 5 - 6 | 616 | 63 | ||
| AY523555.1 | - | 4 - 5 | 587 | 63 | ||
| BC016261.1 | - | 4 - 5 | 584 | 4920560 | 63 | |
| BC021864.1 | - | 5 - 6 | 606 | 5123958 | 63 | |
| BC021949.1 | - | 4 - 5 | 417 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ciapin1
Genomic Map