Assay Details
| Gene Symbol: Mmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008609.3 | NP_032635.1 | 5 - 6 | 1431 | 67 | |
| GenBank mRNA | BC047278.1 | - | 5 - 6 | 1431 | 6390113 | 67 |
| BC057952.1 | - | 5 - 6 | 1001 | 5342023 | 67 | |
| D86332.1 | - | 5 - 6 | 927 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp15
Genomic Map