Assay Details
| Gene Symbol: Cpsf4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164532.1 | NP_001158004.1 | 5 - 6 | 690 | 73 | |
| NM_026682.2 | NP_080958.2 | 5 - 6 | 534 | 73 | ||
| GenBank mRNA | BC016190.1 | - | 5 - 6 | 528 | 4483305 | 73 |
| DV648079.1 | - | 5 - 6 | 661 | 73 | ||
| DV659633.1 | - | 5 - 6 | 524 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf4l
Genomic Map