Assay Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080932.2 | NP_001074401.2 | 4 - 5 | 1025 | 146 | |
| GenBank mRNA | AK150439.1 | - | 4 - 5 | 679 | 146 | |
| EU882160.1 | - | 4 - 5 | 1025 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxk2
Genomic Map