Assay Details
| Gene Symbol: Fn3krp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181420.3 | NP_852085.2 | 2 - 3 | 386 | 82 | |
| GenBank mRNA | AY360466.1 | - | 2 - 3 | 344 | 82 | |
| BC032265.1 | - | 2 - 3 | 332 | 4015688 | 82 | |
More Information
Set Membership: |
|

Fn3krp
Genomic Map