Assay Details
| Gene Symbol: Rnasek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173742.3 | NP_776103.1 | 1 - 2 | 197 | 94 | |
| GenBank mRNA | AK003548.1 | - | 1 - 2 | 173 | 94 | |
| AK009592.1 | - | 1 - 2 | 192 | 94 | ||
| AK018709.1 | - | 1 - 2 | 146 | 94 | ||
| AK171975.1 | - | 1 - 2 | 146 | 94 | ||
| BC021602.1 | - | 1 - 2 | 125 | 5102170 | 94 | |
| BC021603.1 | - | 1 - 2 | 125 | 5102465 | 94 | |
| BC024417.1 | - | 1 - 2 | 155 | 5065541 | 94 | |
| CJ181610.1 | - | 1 - 2 | 197 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnasek
Genomic Map