Assay Details
| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029231.4 | NP_083507.3 | 4 - 5 | 590 | 55 | |
| GenBank mRNA | AK005027.1 | - | 4 - 5 | 588 | 55 | |
| AK128966.1 | - | 4 - 5 | 590 | 55 | ||
| AK217016.1 | - | 1 - 2 | 163 | 55 | ||
| BC090620.1 | - | 4 - 5 | 584 | 30699968 | 55 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pelp1
Genomic Map