Assay Details
| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029231.4 | NP_083507.3 | 8 - 9 | 996 | 75 | |
| GenBank mRNA | AK005027.1 | - | 8 - 9 | 994 | 75 | |
| AK128966.1 | - | 8 - 9 | 996 | 75 | ||
| BC057987.1 | - | 2 - 3 | 224 | 75 | ||
| BC090620.1 | - | 8 - 9 | 990 | 30699968 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pelp1
Genomic Map