Assay Details
| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007440.4 | NP_031466.2 | 6 - 7 | 875 | 68 | |
| GenBank mRNA | AK036898.1 | - | 6 - 7 | 873 | 68 | |
| AK036960.1 | - | 6 - 7 | 831 | 68 | ||
| AK079232.1 | - | 6 - 7 | 876 | 68 | ||
| AK087724.1 | - | 6 - 7 | 876 | 68 | ||
| AK172216.1 | - | 6 - 7 | 873 | 68 | ||
| AK172488.1 | - | 6 - 7 | 876 | 68 | ||
| BC152329.1 | - | 6 - 7 | 880 | 40141477 | 68 | |
| BC152403.1 | - | 6 - 7 | 880 | 40141482 | 68 | |
| S80446.1 | - | 6 - 7 | 887 | 68 | ||
| U04334.1 | - | 6 - 7 | 806 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Alox12
Genomic Map