Assay Details
| Gene Symbol: C1qbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007573.2 | NP_031599.2 | 1 - 2 | 307 | 93 | |
| GenBank mRNA | AF300619.1 | - | 1 - 2 | 246 | 93 | |
| AJ001101.1 | - | 1 - 2 | 252 | 93 | ||
| AK010601.1 | - | 1 - 2 | 245 | 93 | ||
| AK010746.1 | - | 1 - 2 | 271 | 93 | ||
| AK011868.1 | - | 1 - 2 | 298 | 93 | ||
| AK169764.1 | - | 1 - 2 | 290 | 93 | ||
| BC038075.1 | - | 1 - 2 | 291 | 3154276 | 93 | |
| BY275303.1 | - | 1 - 2 | 308 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C1qbp
Genomic Map