Assay Details
| Gene Symbol: C1qbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007573.2 | NP_031599.2 | 3 - 4 | 548 | 100 | |
| GenBank mRNA | AF300619.1 | - | 3 - 4 | 487 | 100 | |
| AK010601.1 | - | 3 - 4 | 486 | 100 | ||
| AK010746.1 | - | 3 - 4 | 512 | 100 | ||
| AK011868.1 | - | 3 - 4 | 539 | 100 | ||
| AK169764.1 | - | 3 - 4 | 531 | 100 | ||
| BC038075.1 | - | 3 - 4 | 532 | 3154276 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

C1qbp
Genomic Map