Assay Details
| Gene Symbol: Spns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029932.3 | NP_084208.1 | 7 - 8 | 977 | 74 | |
| GenBank mRNA | AK020683.1 | - | 7 - 8 | 978 | 74 | |
| AK041376.1 | - | 6 - 7 | 967 | 74 | ||
| AK156479.1 | - | 7 - 8 | 1241 | 74 | ||
| AK156781.1 | - | 7 - 8 | 969 | 74 | ||
| BC150778.1 | - | 7 - 8 | 960 | 9087689 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spns3
Genomic Map