Assay Details
| Gene Symbol: Mpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016695.3 | NP_057904.1 | 2 - 3 | 159 | 63 | |
| GenBank mRNA | AF162685.1 | - | 2 - 3 | 146 | 63 | |
| AK161577.1 | - | 2 - 3 | 152 | 63 | ||
| AK172253.1 | - | 2 - 3 | 144 | 63 | ||
| BC053026.1 | - | 2 - 3 | 159 | 5709543 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF162685.1, AK161577.1, AK172253.1, BC053026.1, NM_016695.3 transcripts.
Set Membership: |
|

Mpp2
Genomic Map