Assay Details
| Gene Symbol: Aspa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023113.5 | NP_075602.2 | 1 - 2 | 383 | 82 | |
| GenBank mRNA | AF212998.1 | - | 1 - 2 | 233 | 82 | |
| AI527854.1 | - | 1 - 2 | 383 | 82 | ||
| AK002859.1 | - | 1 - 2 | 381 | 82 | ||
| AK008354.1 | - | 1 - 2 | 386 | 82 | ||
| AK035910.1 | - | 1 - 2 | 84 | 82 | ||
| BC024934.1 | - | 1 - 2 | 347 | 4012726 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aspa
Genomic Map