Assay Details
| Gene Symbol: Hdac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077696.1 | NP_001071164.1 | 10 - 11 | 1507 | 64 | |
| NM_001284248.1 | NP_001271177.1 | 12 - 13 | 2019 | 64 | ||
| NM_001284249.1 | NP_001271178.1 | 11 - 12 | 1616 | 64 | ||
| NM_001284250.1 | NP_001271179.1 | 10 - 11 | 1504 | 64 | ||
| NM_010412.3 | NP_034542.3 | 10 - 11 | 1504 | 64 | ||
| GenBank mRNA | AF006602.2 | - | 9 - 10 | 1220 | 64 | |
| AF207748.1 | - | 9 - 10 | 1139 | 64 | ||
| AK122325.2 | - | 5 - 6 | 543 | 64 | ||
| AK139202.1 | - | 11 - 12 | 1590 | 64 | ||
| AK145972.1 | - | 10 - 11 | 1470 | 64 | ||
| AK146483.1 | - | 10 - 11 | 1443 | 64 | ||
| AK155335.1 | - | 10 - 11 | 1478 | 64 | ||
| AK191268.1 | - | 1 - 2 | 70 | 64 | ||
| BC023309.1 | - | 1 - 2 | 57 | 64 | ||
| BC049926.1 | - | 10 - 11 | 1440 | 64 | ||
| BC058554.1 | - | 12 - 13 | 1971 | 64 | ||
| BC060609.1 | - | 9 - 10 | 1298 | 5720438 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdac5
Genomic Map