Assay Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252193.1 | NP_001239122.1 | 4 - 5 | 940 | 76 | |
| NM_025275.4 | NP_079551.3 | 5 - 6 | 759 | 76 | ||
| GenBank mRNA | AJ879914.1 | - | 2 - 3 | 456 | 76 | |
| AK075748.1 | - | 5 - 6 | 699 | 76 | ||
| AK143908.1 | - | 5 - 6 | 724 | 76 | ||
| BC025087.1 | - | 4 - 5 | 934 | 2649126 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075748.1 transcripts.
Set Membership: |
|

Amz2
Genomic Map