| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100017230.mm.1 |
NM_001252193.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017230.mm.1 |
NM_001252193.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017230.mm.1 |
NM_025275.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017230.mm.1 |
NM_025275.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017230.mm.1 |
TC1100001830.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017229.mm.1 |
NM_001252193.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017229.mm.1 |
NM_001252193.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017229.mm.1 |
NM_025275.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017229.mm.1 |
NM_025275.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017229.mm.1 |
TC1100001830.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017231.mm.1 |
NM_001252193.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017231.mm.1 |
NM_001252193.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017231.mm.1 |
NM_025275.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017231.mm.1 |
NM_025275.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017231.mm.1 |
TC1100001830.mm.1 |
MTA TC |