Assay Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001986.2 | NP_001001986.2 | 5 - 6 | 914 | 91 | |
| NM_001134300.2 | NP_001127772.1 | 8 - 9 | 1699 | 91 | ||
| GenBank mRNA | AK139459.1 | - | 5 - 6 | 914 | 91 | |
| AK147524.1 | - | 8 - 9 | 1701 | 91 | ||
| AK157863.1 | - | 5 - 6 | 895 | 91 | ||
| BC075648.1 | - | 7 - 8 | 1110 | 30620653 | 91 | |
| BC079552.1 | - | 5 - 6 | 856 | 91 | ||
| BC094234.1 | - | 4 - 5 | 765 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nol4l
Genomic Map