Assay Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134300.2 | NP_001127772.1 | 1 - 2 | 529 | 78 | |
| GenBank mRNA | AK147524.1 | - | 1 - 2 | 531 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nol4l
Genomic Map