Assay Details
| Gene Symbol: Tk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271729.1 | NP_001258658.1 | 2 - 3 | 412 | 63 | |
| NM_009387.2 | NP_033413.2 | 3 - 4 | 444 | 63 | ||
| GenBank mRNA | AK085188.1 | - | 3 - 4 | 293 | 63 | |
| AK087988.1 | - | 3 - 4 | 247 | 63 | ||
| AK139244.1 | - | 3 - 4 | 264 | 63 | ||
| BC012215.1 | - | 3 - 4 | 248 | 3969287 | 63 | |
| BC091758.1 | - | 3 - 4 | 277 | 4978985 | 63 | |
| CF732600.1 | - | 2 - 3 | 227 | 63 | ||
| M11945.1 | - | 3 - 4 | 241 | 63 | ||
| M19438.1 | - | 2 - 3 | 134 | 63 | ||
| S57244.1 | - | 3 - 4 | 241 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tk1
Genomic Map