Assay Details
| Gene Symbol: Tk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271729.1 | NP_001258658.1 | 3 - 4 | 510 | 77 | |
| NM_009387.2 | NP_033413.2 | 4 - 5 | 542 | 77 | ||
| GenBank mRNA | AK085188.1 | - | 4 - 5 | 391 | 77 | |
| AK087988.1 | - | 4 - 5 | 345 | 77 | ||
| AK139244.1 | - | 4 - 5 | 362 | 77 | ||
| BC012215.1 | - | 4 - 5 | 346 | 3969287 | 77 | |
| BC091758.1 | - | 4 - 5 | 375 | 4978985 | 77 | |
| CF732600.1 | - | 3 - 4 | 325 | 77 | ||
| M11945.1 | - | 4 - 5 | 339 | 77 | ||
| M19438.1 | - | 3 - 4 | 232 | 77 | ||
| S57244.1 | - | 4 - 5 | 339 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tk1
Genomic Map