Assay Details
| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC011503.1 | - | 4 - 5 | 1596 | 113 | |
| BC117737.1 | - | 4 - 5 | 927 | 113 | ||
| EF527404.1 | - | 5 - 6 | 1281 | 113 | ||
| EF527405.1 | - | 5 - 6 | 1029 | 113 | ||
More Information
Set Membership: |
|

Jmjd6
Genomic Map